3,5-Dichlor-4'-fluorbenzhydrol

Lieferant: FLUOROCHEM

022511-1G
FLUO022511-1GEA 1010 EUR
FLUO022511-1G
3,5-Dichlor-4'-fluorbenzhydrol
3,5-Dichlor-4'-fluorbenzhydrol
Formel: C₁₃H₉Cl₂FO
MDL: MFCD06201337
CAS-Nummer: 844683-76-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR