4-Fluor-3',4'-dimethoxybenzhydrol

Lieferant: FLUOROCHEM

022514-1G
FLUO022514-1GEA 1010 EUR
FLUO022514-1G
4-Fluor-3',4'-dimethoxybenzhydrol
4-Fluor-3',4'-dimethoxybenzhydrol
Formel: C₁₅H₁₅FO₃
CAS-Nummer: 844856-32-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR