4-(Dimethylamino)-4'-fluorbenzhydrol

Lieferant: FLUOROCHEM

022515-1G
FLUO022515-1GEA 1010 EUR
FLUO022515-1G
4-(Dimethylamino)-4'-fluorbenzhydrol
4-(Dimethylamino)-4'-fluorbenzhydrol
Formel: C₁₅H₁₆FNO
CAS-Nummer: 39768-80-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR