3,5-Dimethyl-4'-fluorbenzhydrol

Lieferant: FLUOROCHEM

022516-1G
FLUO022516-1GEA 1010 EUR
FLUO022516-1G
3,5-Dimethyl-4'-fluorbenzhydrol
3,5-Dimethyl-4'-fluorbenzhydrol
Formel: C₁₅H₁₅FO
CAS-Nummer: 844856-33-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR