4-Ethyl-4'-fluorbenzhydrol
Lieferant: FLUOROCHEM
022517-1G
FLUO022517-1GEA
1200
EUR
FLUO022517-1G
4-Ethyl-4'-fluorbenzhydrol
4-Ethyl-4'-fluorbenzhydrol
Formel:
C₁₅H₁₅FO |
MDL:
MFCD06201343 CAS-Nummer: 229962-77-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...