3,4'-Difluorbenzhydrol

Lieferant: FLUOROCHEM

022519-1G
FLUO022519-1GEA 1010 EUR
FLUO022519-1G
3,4'-Difluorbenzhydrol
3,4'-Difluorbenzhydrol
Formel: C₁₃H₁₀F₂O
MDL: MFCD06201345
CAS-Nummer: 190444-04-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR