4-Fluor-4'-(methylthio)benzhydrol

Lieferant: FLUOROCHEM

022522-1G
FLUO022522-1GEA 1010 EUR
FLUO022522-1G
4-Fluor-4'-(methylthio)benzhydrol
4-Fluor-4'-(methylthio)benzhydrol
Formel: C₁₄H₁₃FOS
CAS-Nummer: 1510-48-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR