3,5-Dichlor-3'-methylbenzhydrol
Lieferant: FLUOROCHEM
022523-1G
FLUO022523-1GEA
1010
EUR
FLUO022523-1G
3,5-Dichlor-3'-methylbenzhydrol
3,5-Dichlor-3'-methylbenzhydrol
Formel:
C₁₄H₁₂Cl₂O |
MDL:
MFCD06201349 CAS-Nummer: 844856-36-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...