4-Brom-4'-methylbiphenyl

Lieferant: FLUOROCHEM

Synonyme: 4-Bromo-4'-methyl-1,1'-biphenyl

022566-1G
FLUO022566-1GEA 84.9 EUR
FLUO022566-1G
4-Brom-4'-methylbiphenyl
4-Brom-4'-methylbiphenyl
Formel: C₁₃H₁₁Br
Molecular Weight: 247,14 g/mol
MDL: MFCD00466236
CAS-Nummer: 50670-49-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR