3-Brom-4'-fluorbiphenyl
Lieferant: FLUOROCHEM
Synonyme:
3-Bromo-4'-Fluoro-1,1'-biphenyl
022588-1G
FLUO022588-1GEA
336
EUR
FLUO022588-1G
3-Brom-4'-fluorbiphenyl
3-Brom-4'-fluorbiphenyl
Formel:
C₁₂H₈BrF Molecular Weight: 251,0943 g/mol |
MDL:
MFCD04038228 CAS-Nummer: 10540-35-9 EINECS: 206-909-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...