2-Methoxy-p-toluylsäure

Lieferant: FLUOROCHEM

Synonyme: 2-Methoxy-4-methylbenzoesäure

022645-1G
FLUO022645-1GEA 36.9 EUR
FLUO022645-1G
2-Methoxy-p-toluylsäure
2-Methoxy-p-toluylsäure
Formel: C₉H₁₀O₃
Molecular Weight: 166,18 g/mol
MDL: MFCD00020380
CAS-Nummer: 704-45-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR