4'-Chlor-3'-fluorbiphenyl-3-carbonsäure
Lieferant: FLUOROCHEM
022647-1G
FLUO022647-1GEA
0
EUR
FLUO022647-1G
4'-Chlor-3'-fluorbiphenyl-3-carbonsäure
4'-Chlor-3'-fluorbiphenyl-3-carbonsäure
Formel:
C₁₃H₈ClFO₂ |
MDL:
MFCD06201433 CAS-Nummer: 844878-88-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...