3',5'-Dimethylbiphenyl-3-carbonsäure

Lieferant: FLUOROCHEM

022653-1G
FLUO022653-1GEA 0 EUR
FLUO022653-1G
3',5'-Dimethylbiphenyl-3-carbonsäure
3',5'-Dimethylbiphenyl-3-carbonsäure
Formel: C₁₅H₁₄O₂
MDL: MFCD06201437
CAS-Nummer: 177734-84-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR