6-Methoxy-2-naphthoesäure
Lieferant: FLUOROCHEM
022658-1G
FLUO022658-1GEA
42.9
EUR
FLUO022658-1G
6-Methoxy-2-naphthoesäure
6-Methoxy-2-naphthoesäure
Formel:
C₁₂H₁₀O₃ |
MDL:
MFCD00092750 CAS-Nummer: 2471-70-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...