4-Brom-3'-methoxybenzophenon

Lieferant: FLUOROCHEM

022662-1G
FLUO022662-1GEA 830 EUR
FLUO022662-1G
4-Brom-3'-methoxybenzophenon
4-Brom-3'-methoxybenzophenon
Formel: C₁₄H₁₁BrO₂
MDL: MFCD01311563
CAS-Nummer: 151239-47-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR