4-Brom-4'-methoxybenzophenon
Lieferant: FLUOROCHEM
Synonyme:
(4-Bromophenyl)(4-methoxyphenyl)methanone
022663-1G
FLUO022663-1GEA
461
EUR
FLUO022663-1G
4-Brom-4'-methoxybenzophenon
4-Brom-4'-methoxybenzophenon
Formel:
C₁₄H₁₁BrO₂ Molecular Weight: 291,14 g/mol |
MDL:
MFCD00228667 CAS-Nummer: 54118-75-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...