4-Brom-4'-(methylthio)benzophenon

Lieferant: FLUOROCHEM

022664-1G
FLUO022664-1GEA 830 EUR
FLUO022664-1G
4-Brom-4'-(methylthio)benzophenon
4-Brom-4'-(methylthio)benzophenon
Formel: C₁₄H₁₁BrOS
MDL: MFCD06201441
CAS-Nummer: 197439-28-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR