4-Brom-4'-tert-butylbenzophenon

Lieferant: FLUOROCHEM

022666-1G
FLUO022666-1GEA 28.2 EUR
FLUO022666-1G
4-Brom-4'-tert-butylbenzophenon
4-Brom-4'-tert-butylbenzophenon
Formel: C₁₇H₁₇BrO
MDL: MFCD06201443
CAS-Nummer: 91404-26-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR