4-Brom-3',4'-difluorbenzophenon

Lieferant: FLUOROCHEM

022668-1G
FLUO022668-1GEA 830 EUR
FLUO022668-1G
4-Brom-3',4'-difluorbenzophenon
4-Brom-3',4'-difluorbenzophenon
Formel: C₁₃H₇BrF₂O
MDL: MFCD06201445
CAS-Nummer: 844878-99-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR