4-Brom-3',5'-difluorbenzophenon
Lieferant: FLUOROCHEM
Synonyme:
(4-Bromophenyl)(3,5-difluorophenyl)methanone
022669-1G
FLUO022669-1GEA
830
EUR
FLUO022669-1G
4-Brom-3',5'-difluorbenzophenon
4-Brom-3',5'-difluorbenzophenon
Formel:
C₁₃H₇BrF₂O Molecular Weight: 297,949 g/mol |
MDL:
MFCD06201446 CAS-Nummer: 844879-03-4 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...