4-Brom-3',5'-dichlorbenzophenon

Lieferant: FLUOROCHEM

022671-1G
FLUO022671-1GEA 830 EUR
FLUO022671-1G
4-Brom-3',5'-dichlorbenzophenon
4-Brom-3',5'-dichlorbenzophenon
Formel: C₁₃H₇BrCl₂O
MDL: MFCD06201448
CAS-Nummer: 844879-09-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR