4-Brom-3'-chlor-5'-fluorbenzophenon
Lieferant: FLUOROCHEM
022672-1G
FLUO022672-1GEA
830
EUR
FLUO022672-1G
4-Brom-3'-chlor-5'-fluorbenzophenon
4-Brom-3'-chlor-5'-fluorbenzophenon
Formel:
C₁₃H₇BrClFO |
MDL:
MFCD06201449 CAS-Nummer: 844879-12-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...