4-Brom-3',4'-dimethoxybenzophenon

Lieferant: FLUOROCHEM

022673-1G
FLUO022673-1GEA 830 EUR
FLUO022673-1G
4-Brom-3',4'-dimethoxybenzophenon
4-Brom-3',4'-dimethoxybenzophenon
Formel: C₁₅H₁₃BrO₃
MDL: MFCD02127292
CAS-Nummer: 116412-90-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR