4-Brom-3',5'-dimethyl-4'-methoxybenzophenon

Lieferant: FLUOROCHEM

022674-1G
FLUO022674-1GEA 882 EUR
FLUO022674-1G
4-Brom-3',5'-dimethyl-4'-methoxybenzophenon
4-Brom-3',5'-dimethyl-4'-methoxybenzophenon
Formel: C₁₆H₁₅BrO₂
MDL: MFCD06201450
CAS-Nummer: 844879-14-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR