4-Brom-3',4'-dimethylbenzophenon

Lieferant: FLUOROCHEM

022675-1G
FLUO022675-1GEA 830 EUR
FLUO022675-1G
4-Brom-3',4'-dimethylbenzophenon
4-Brom-3',4'-dimethylbenzophenon
Formel: C₁₅H₁₃BrO
MDL: MFCD00625700
CAS-Nummer: 300383-01-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR