4-Brom-3',5'-dimethylbenzophenon

Lieferant: FLUOROCHEM

022676-1G
FLUO022676-1GEA 830 EUR
FLUO022676-1G
4-Brom-3',5'-dimethylbenzophenon
4-Brom-3',5'-dimethylbenzophenon
Formel: C₁₅H₁₃BrO
MDL: MFCD06201451
CAS-Nummer: 844879-16-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR