4-Brom-4'-(ethylthio)benzophenon

Lieferant: FLUOROCHEM

022677-1G
FLUO022677-1GEA 882 EUR
FLUO022677-1G
4-Brom-4'-(ethylthio)benzophenon
4-Brom-4'-(ethylthio)benzophenon
Formel: C₁₅H₁₃BrOS
MDL: MFCD06201452
CAS-Nummer: 951885-47-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR