4-Brom-3'-fluor-4'-methoxybenzophenon

Lieferant: FLUOROCHEM

022678-1G
FLUO022678-1GEA 830 EUR
FLUO022678-1G
4-Brom-3'-fluor-4'-methoxybenzophenon
4-Brom-3'-fluor-4'-methoxybenzophenon
Formel: C₁₄H₁₀BrFO₂
MDL: MFCD06201453
CAS-Nummer: 161581-93-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR