3-Brom-3'-methoxybenzophenon

Lieferant: FLUOROCHEM

022684-1G
FLUO022684-1GEA 830 EUR
FLUO022684-1G
3-Brom-3'-methoxybenzophenon
3-Brom-3'-methoxybenzophenon
Formel: C₁₄H₁₁BrO₂
MDL: MFCD01311561
CAS-Nummer: 750633-66-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR