3-Brom-4'-methoxybenzophenon

Lieferant: FLUOROCHEM

022685-1G
FLUO022685-1GEA 29.3 EUR
FLUO022685-1G
3-Brom-4'-methoxybenzophenon
3-Brom-4'-methoxybenzophenon
Formel: C₁₄H₁₁BrO₂
MDL: MFCD01311588
CAS-Nummer: 54118-76-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR