3-Brom-4'-(methylthio)benzophenon
Lieferant: FLUOROCHEM
022686-1G
FLUO022686-1GEA
830
EUR
FLUO022686-1G
3-Brom-4'-(methylthio)benzophenon
3-Brom-4'-(methylthio)benzophenon
Formel:
C₁₄H₁₁BrOS |
MDL:
MFCD06201459 CAS-Nummer: 197439-23-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...