3-Brom-4'-tert-butylbenzophenon

Lieferant: FLUOROCHEM

022688-1G
FLUO022688-1GEA 830 EUR
FLUO022688-1G
3-Brom-4'-tert-butylbenzophenon
3-Brom-4'-tert-butylbenzophenon
Formel: C₁₇H₁₇BrO
MDL: MFCD06201461
CAS-Nummer: 132885-82-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR