3-Brom-4'-butylbenzophenon

Lieferant: FLUOROCHEM

022691-1G
FLUO022691-1GEA 882 EUR
FLUO022691-1G
3-Brom-4'-butylbenzophenon
3-Brom-4'-butylbenzophenon
Formel: C₁₇H₁₇BrO
MDL: MFCD06201464
CAS-Nummer: 844879-33-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR