3-Brom-3',4'-difluorbenzophenon

Lieferant: FLUOROCHEM

022692-1G
FLUO022692-1GEA 830 EUR
FLUO022692-1G
3-Brom-3',4'-difluorbenzophenon
3-Brom-3',4'-difluorbenzophenon
Formel: C₁₃H₇BrF₂O
MDL: MFCD06201465
CAS-Nummer: 844879-35-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR