3-Brom-3',5'-difluorbenzophenon

Lieferant: FLUOROCHEM

022693-1G
FLUO022693-1GEA 830 EUR
FLUO022693-1G
3-Brom-3',5'-difluorbenzophenon
3-Brom-3',5'-difluorbenzophenon
Formel: C₁₃H₇BrF₂O
MDL: MFCD06201466
CAS-Nummer: 844879-37-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR