3-Brom-3',4'-dichlorbenzophenon

Lieferant: FLUOROCHEM

022694-1G
FLUO022694-1GEA 830 EUR
FLUO022694-1G
3-Brom-3',4'-dichlorbenzophenon
3-Brom-3',4'-dichlorbenzophenon
Formel: C₁₃H₇BrCl₂O
MDL: MFCD06201467
CAS-Nummer: 844879-39-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR