3-Brom-3',5'-dichlorbenzophenon

Lieferant: FLUOROCHEM

022695-1G
FLUO022695-1GEA 830 EUR
FLUO022695-1G
3-Brom-3',5'-dichlorbenzophenon
3-Brom-3',5'-dichlorbenzophenon
Formel: C₁₃H₇BrCl₂O
MDL: MFCD06201468
CAS-Nummer: 844879-42-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR