3-Brom-3'-chlor-5'-fluorbenzophenon
Lieferant: FLUOROCHEM
022696-1G
FLUO022696-1GEA
830
EUR
FLUO022696-1G
3-Brom-3'-chlor-5'-fluorbenzophenon
3-Brom-3'-chlor-5'-fluorbenzophenon
Formel:
C₁₃H₇BrClFO |
MDL:
MFCD06201469 CAS-Nummer: 844879-45-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...