3-Brom-3',4'-dimethoxybenzophenon

Lieferant: FLUOROCHEM

022697-1G
FLUO022697-1GEA 830 EUR
FLUO022697-1G
3-Brom-3',4'-dimethoxybenzophenon
3-Brom-3',4'-dimethoxybenzophenon
Formel: C₁₅H₁₃BrO₃
MDL: MFCD02127297
CAS-Nummer: 681469-97-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR