3-Brom-3',5'-dimethyl-4'-methoxybenzophenon

Lieferant: FLUOROCHEM

022698-1G
FLUO022698-1GEA 672 EUR
FLUO022698-1G
3-Brom-3',5'-dimethyl-4'-methoxybenzophenon
3-Brom-3',5'-dimethyl-4'-methoxybenzophenon
Formel: C₁₆H₁₅BrO₂
MDL: MFCD06201470
CAS-Nummer: 844879-47-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR