3-Brom-3',4'-dimethylbenzophenon
Lieferant: FLUOROCHEM
022699-1G
FLUO022699-1GEA
830
EUR
FLUO022699-1G
3-Brom-3',4'-dimethylbenzophenon
3-Brom-3',4'-dimethylbenzophenon
Formel:
C₁₅H₁₃BrO |
MDL:
MFCD06201471 CAS-Nummer: 844879-49-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...