3-Brom-3',5'-dimethylbenzophenon
Lieferant: FLUOROCHEM
022700-1G
FLUO022700-1GEA
830
EUR
FLUO022700-1G
3-Brom-3',5'-dimethylbenzophenon
3-Brom-3',5'-dimethylbenzophenon
Formel:
C₁₅H₁₃BrO |
MDL:
MFCD06201472 CAS-Nummer: 844879-51-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...