3-Brom-3'-fluor-4'-methoxybenzophenon
Lieferant: FLUOROCHEM
022702-1G
FLUO022702-1GEA
830
EUR
FLUO022702-1G
3-Brom-3'-fluor-4'-methoxybenzophenon
3-Brom-3'-fluor-4'-methoxybenzophenon
Formel:
C₁₄H₁₀BrFO₂ |
MDL:
MFCD06201474 CAS-Nummer: 844879-54-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...