3-Chlor-4'-methoxybenzophenon

Lieferant: FLUOROCHEM

Synonyme: (3-Chlorophenyl)(4-methoxyphenyl)methanone

022709-1G
FLUO022709-1GEA 461 EUR
FLUO022709-1G
3-Chlor-4'-methoxybenzophenon
3-Chlor-4'-methoxybenzophenon
Formel: C₁₄H₁₁ClO₂
Molecular Weight: 246,69 g/mol
MDL: MFCD01311591
CAS-Nummer: 13389-51-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR