3-Chlor-4'-(methylthio)benzophenon
Lieferant: FLUOROCHEM
022710-1G
FLUO022710-1GEA
830
EUR
FLUO022710-1G
3-Chlor-4'-(methylthio)benzophenon
3-Chlor-4'-(methylthio)benzophenon
Formel:
C₁₄H₁₁ClOS |
MDL:
MFCD06201479 CAS-Nummer: 197438-99-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...