4-tert-Butyl-3'-chlorbenzophenon

Lieferant: FLUOROCHEM

022712-1G
FLUO022712-1GEA 882 EUR
FLUO022712-1G
4-tert-Butyl-3'-chlorbenzophenon
4-tert-Butyl-3'-chlorbenzophenon
Formel: C₁₇H₁₇ClO
MDL: MFCD06201481
CAS-Nummer: 93977-28-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR