4-Butyl-3'-chlorbenzophenon

Lieferant: FLUOROCHEM

022715-1G
FLUO022715-1GEA 882 EUR
FLUO022715-1G
4-Butyl-3'-chlorbenzophenon
4-Butyl-3'-chlorbenzophenon
Formel: C₁₇H₁₇ClO
MDL: MFCD06201484
CAS-Nummer: 844884-93-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR