3-Chlor-3',4'-difluorbenzophenon
Lieferant: FLUOROCHEM
022716-1G
FLUO022716-1GEA
830
EUR
FLUO022716-1G
3-Chlor-3',4'-difluorbenzophenon
3-Chlor-3',4'-difluorbenzophenon
Formel:
C₁₃H₇ClF₂O |
MDL:
MFCD06201485 CAS-Nummer: 844884-94-2 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...