3,3'-Dichlor-5-fluorbenzophenon
Lieferant: FLUOROCHEM
022719-1G
FLUO022719-1GEA
830
EUR
FLUO022719-1G
3,3'-Dichlor-5-fluorbenzophenon
3,3'-Dichlor-5-fluorbenzophenon
Formel:
C₁₃H₇Cl₂FO |
MDL:
MFCD06201488 CAS-Nummer: 844884-96-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...