4-Chlor-3',4'-dimethoxybenzophenon

Lieferant: FLUOROCHEM

Synonyme: (4-Chlorophenyl)(3,4-dimethoxyphenyl)methanone

022720-1G
FLUO022720-1GEA 830 EUR
FLUO022720-1G
4-Chlor-3',4'-dimethoxybenzophenon
4-Chlor-3',4'-dimethoxybenzophenon
Formel: C₁₅H₁₃ClO₃
Molecular Weight: 276,7149 g/mol
MDL: MFCD02127307
CAS-Nummer: 116412-83-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR